Assay Details
| Gene Symbol: UbcE2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132210.3 | NP_572438.1 | 2 | 2023 | 113 | |
| NM_167145.2 | NP_727233.1 | 2 | 2160 | 113 | ||
| NM_167146.2 | NP_727234.1 | 2 | 2059 | 113 | ||
| GenBank mRNA | AY061237.1 | - | 2 | 2002 | 113 | |
More Information
Set Membership: |
|

UbcE2H
Genomic Map