Assay Details
| Gene Symbol: MTPAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272200.1 | NP_001259129.1 | 4 | 2128 | 87 | |
| NM_130548.3 | NP_569904.1 | 3 | 2204 | 87 | ||
| NM_206602.2 | NP_996325.1 | 2 | 1259 | 87 | ||
| GenBank mRNA | BT132966.1 | - | 3 | 2204 | 87 | |
More Information
Set Membership: |
|

MTPAP
Genomic Map