Assay Details
| Gene Symbol: par-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014539.3 | NP_001014539.1 | 2 - 3 | 1660 | 113 | |
| GenBank mRNA | AY128423.1 | - | 2 - 3 | 1626 | 113 | |
More Information
Set Membership: |
|

par-1
Genomic Map