Assay Details
| Gene Symbol: Cpr30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135451.3 | NP_609295.1 | 2 | 482 | 63 | |
| GenBank mRNA | BT100143.4 | - | 1 | 413 | 63 | |
| KX531620.1 | - | 1 | 413 | 63 | ||
More Information
Set Membership: |
|

Cpr30B
Genomic Map