Assay Details
| Gene Symbol: nompB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078889.5 | NP_523613.4 | 2 - 3 | 393 | 101 | |
| NM_165385.4 | NP_724347.3 | 2 - 3 | 393 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nompB| Gene Symbol: nompB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078889.5 | NP_523613.4 | 2 - 3 | 393 | 101 | |
| NM_165385.4 | NP_724347.3 | 2 - 3 | 393 | 101 | ||
Set Membership: |
|