Assay Details
| Gene Symbol: nompB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078889.5 | NP_523613.4 | 4 - 5 | 1363 | 153 | |
| NM_165385.4 | NP_724347.3 | 4 - 5 | 1342 | 153 | ||
More Information
Set Membership: |
|

nompB
Genomic Map