Assay Details
| Gene Symbol: CG9667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300295.1 | NP_001287224.1 | 2 - 3 | 188 | 62 | |
| NM_141511.3 | NP_649768.2 | 3 - 4 | 233 | 62 | ||
| GenBank mRNA | AY084151.1 | - | 3 - 4 | 209 | 62 | |
More Information
Set Membership: |
|

CG9667
Genomic Map