Assay Details
| Gene Symbol: Sxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031892.2 | NP_001027063.1 | 6 | 1228 | 103 | |
| NM_001031894.1 | NP_001027065.1 | 7 | 1567 | 103 | ||
| NM_001031895.1 | NP_001027066.1 | 8 | 1843 | 103 | ||
| NM_001169216.2 | NP_001162687.1 | 9 | 1825 | 103 | ||
| NM_001169218.3 | NP_001162689.1 | 7 | 1513 | 103 | ||
| NM_001169219.1 | NP_001162690.1 | 7 | 1623 | 103 | ||
| NM_001272376.1 | NP_001259305.1 | 8 | 1787 | 103 | ||
| NM_001272377.1 | NP_001259306.1 | 8 | 1781 | 103 | ||
| NM_001316622.1 | NP_001303551.1 | 7 | 1489 | 103 | ||
| NM_080052.4 | NP_524791.3 | 6 | 1228 | 103 | ||
| NM_167110.3 | NP_727160.2 | 7 | 1599 | 103 | ||
| NM_167112.2 | NP_727162.2 | 7 | 1567 | 103 | ||
| NM_167113.3 | NP_727163.2 | 8 | 1333 | 103 | ||
| NM_167115.3 | NP_727165.2 | 8 | 1403 | 103 | ||
| NM_167116.2 | NP_727166.2 | 8 | 1697 | 103 | ||
| NM_167117.2 | NP_727167.2 | 8 | 1733 | 103 | ||
| NM_167118.2 | NP_727168.1 | 8 | 1733 | 103 | ||
| GenBank mRNA | BT125904.1 | - | 6 | 1220 | 103 | |
| M23635.1 | - | 8 | 1782 | 103 | ||
| M23636.1 | - | 7 | 1624 | 103 | ||
| M59447.1 | - | 7 | 1567 | 103 | ||
More Information
Set Membership: |
|

Sxl
Genomic Map