Assay Details
| Gene Symbol: RpL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110959.3 | NP_001104429.2 | 1 - 2 | 206 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001110959.3 transcripts.
Set Membership: |
|

RpL15
Genomic Map