Assay Details
| Gene Symbol: RpL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042922.3 | NP_001036387.1 | 3 - 4 | 810 | 80 | |
| NM_001042923.3 | NP_001036388.1 | 4 - 5 | 755 | 80 | ||
| NM_001042925.3 | NP_001036390.2 | 4 - 5 | 792 | 80 | ||
| NM_001042926.2 | NP_001036391.2 | 3 - 4 | 1044 | 80 | ||
| GenBank mRNA | AY071305.1 | - | 4 - 5 | 792 | 80 | |
More Information
Set Membership: |
|

RpL5
Genomic Map