Assay Details
| Gene Symbol: Gprk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042973.3 | NP_001036438.1 | 13 - 14 | 1934 | 149 | |
| NM_001169580.2 | NP_001163051.1 | 14 - 15 | 1849 | 149 | ||
| NM_001299206.1 | NP_001286135.1 | 13 - 14 | 1934 | 149 | ||
| GenBank mRNA | BT022770.1 | - | 13 - 14 | 1867 | 149 | |
| BT022801.1 | - | 13 - 14 | 1909 | 149 | ||
More Information
Set Membership: |
|

Gprk1
Genomic Map