Assay Details
| Gene Symbol: Gprk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042973.3 | NP_001036438.1 | 1 - 2 | 291 | 129 | |
| NM_001169580.2 | NP_001163051.1 | 2 - 3 | 206 | 129 | ||
| NM_001299206.1 | NP_001286135.1 | 1 - 2 | 291 | 129 | ||
| GenBank mRNA | BT022770.1 | - | 1 - 2 | 224 | 129 | |
| BT022801.1 | - | 1 - 2 | 266 | 129 | ||
More Information
Set Membership: |
|

Gprk1
Genomic Map