Assay Details
| Gene Symbol: Gprk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042973.3 | NP_001036438.1 | 3 - 4 | 438 | 146 | |
| NM_001169580.2 | NP_001163051.1 | 4 - 5 | 353 | 146 | ||
| NM_001299206.1 | NP_001286135.1 | 3 - 4 | 438 | 146 | ||
| GenBank mRNA | BT022770.1 | - | 3 - 4 | 371 | 146 | |
| BT022801.1 | - | 3 - 4 | 413 | 146 | ||
More Information
Set Membership: |
|

Gprk1
Genomic Map