Assay Details
| Gene Symbol: Gprk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042973.3 | NP_001036438.1 | 7 - 8 | 726 | 122 | |
| NM_001169580.2 | NP_001163051.1 | 8 - 9 | 641 | 122 | ||
| NM_001299206.1 | NP_001286135.1 | 7 - 8 | 726 | 122 | ||
| GenBank mRNA | BT022770.1 | - | 7 - 8 | 659 | 122 | |
| BT022801.1 | - | 7 - 8 | 701 | 122 | ||
More Information
Set Membership: |
|

Gprk1
Genomic Map