Assay Details
| Gene Symbol: cep43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002125.1 | NP_001002125.1 | 13 - 14 | 1093 | 80 | |
| XM_005160662.4 | XP_005160719.1 | 14 - 15 | 1190 | 80 | ||
| GenBank mRNA | BC071450.1 | - | 13 - 14 | 1093 | 80 | |
More Information
Set Membership: |
|

cep43
Genomic Map