Assay Details
| Gene Symbol: cep43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002125.1 | NP_001002125.1 | 17 - 18 | 1390 | 143 | |
| XM_021468391.1 | XP_021324066.1 | 19 - 20 | 1562 | 143 | ||
| GenBank mRNA | BC071450.1 | - | 17 - 18 | 1390 | 143 | |
More Information
Set Membership: |
|

cep43
Genomic Map