Assay Details
| Gene Symbol: cep43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002125.1 | NP_001002125.1 | 9 - 10 | 658 | 96 | |
| XM_005160662.4 | XP_005160719.1 | 9 - 10 | 692 | 96 | ||
| XM_005160663.4 | XP_005160720.1 | 9 - 10 | 692 | 96 | ||
| XM_005160664.4 | XP_005160721.1 | 9 - 10 | 692 | 96 | ||
| XM_017352461.2 | XP_017207950.1 | 9 - 10 | 692 | 96 | ||
| XM_021468390.1 | XP_021324065.1 | 9 - 10 | 692 | 96 | ||
| XM_021468391.1 | XP_021324066.1 | 9 - 10 | 692 | 96 | ||
| XM_021468392.1 | XP_021324067.1 | 9 - 10 | 692 | 96 | ||
| XM_021468393.1 | XP_021324068.1 | 9 - 10 | 692 | 96 | ||
| GenBank mRNA | BC071450.1 | - | 9 - 10 | 658 | 96 | |
More Information
Set Membership: |
|

cep43
Genomic Map