Assay Details
| Gene Symbol: dnajc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003476.2 | NP_001003476.2 | 4 - 5 | 331 | 61 | |
| GenBank mRNA | BC076461.1 | - | 317 | 61 | ||
| BC165133.1 | - | 4 - 5 | 344 | 61 | ||
| EH442214.1 | - | 4 - 5 | 348 | 61 | ||
More Information
Set Membership: |
|

dnajc15
Genomic Map