Assay Details
| Gene Symbol: rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004002.1 | NP_001004002.1 | 6 - 7 | 661 | 101 | |
| GenBank mRNA | BC080219.1 | - | 6 - 7 | 661 | 101 | |
More Information
Set Membership: |
|

rab4b
Genomic Map