Assay Details
| Gene Symbol: fra10ac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006006.2 | NP_001006006.2 | 9 - 10 | 696 | 124 | |
| GenBank mRNA | BC083487.1 | - | 9 - 10 | 671 | 124 | |
More Information
Set Membership: |
|

fra10ac1
Genomic Map