Assay Details
| Gene Symbol: ccdc113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006061.1 | NP_001006061.1 | 4 - 5 | 510 | 89 | |
| GenBank mRNA | BC083263.1 | - | 4 - 5 | 510 | 89 | |
More Information
Set Membership: |
|

ccdc113
Genomic Map