Assay Details
| Gene Symbol: msi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013516.1 | NP_001013534.1 | 10 - 11 | 843 | 96 | |
| GenBank mRNA | AB698465.1 | - | 10 - 11 | 843 | 96 | |
| BC090916.1 | - | 10 - 11 | 843 | 96 | ||
| BC164361.1 | - | 10 - 11 | 835 | 96 | ||
More Information
Set Membership: |
|

msi1b
Genomic Map