Assay Details
| Gene Symbol: msi1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013516.1 | NP_001013534.1 | 8 - 9 | 697 | 73 | |
| GenBank mRNA | AB698464.1 | - | 8 - 9 | 697 | 73 | |
| AB698465.1 | - | 8 - 9 | 697 | 73 | ||
| AB698466.1 | - | 9 - 10 | 732 | 73 | ||
| BC090916.1 | - | 8 - 9 | 697 | 73 | ||
| BC164361.1 | - | 8 - 9 | 689 | 73 | ||
More Information
Set Membership: |
|

msi1b
Genomic Map