Assay Details
| Gene Symbol: rpap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024432.3 | NP_001019603.2 | 7 - 8 | 536 | 92 | |
| NM_001277930.1 | NP_001264859.1 | 7 - 8 | 536 | 92 | ||
| XM_005171173.4 | XP_005171230.1 | 7 - 8 | 559 | 92 | ||
| GenBank mRNA | BC095735.1 | - | 7 - 8 | 522 | 92 | |
More Information
Set Membership: |
|

rpap2
Genomic Map