Assay Details
| Gene Symbol: syngr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320463.1 | NP_001307392.1 | 3 - 4 | 585 | 141 | |
| GenBank mRNA | BC129038.1 | - | 3 - 4 | 585 | 141 | |
| CN320653.1 | - | 3 - 4 | 584 | 141 | ||
More Information
Set Membership: |
|

syngr2a
Genomic Map