Assay Details
| Gene Symbol: coro1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109707.1 | NP_001103177.1 | 6 - 7 | 1163 | 61 | |
| GenBank mRNA | BC152501.1 | - | 6 - 7 | 1163 | 61 | |
More Information
Set Membership: |
|

coro1b
Genomic Map