Assay Details
| Gene Symbol: cct3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173250.1 | NP_775357.1 | 10 - 11 | 1020 | 60 | |
| GenBank mRNA | AF506209.1 | - | 10 - 11 | 1020 | 60 | |
| BC053271.1 | - | 10 - 11 | 1006 | 60 | ||
More Information
Set Membership: |
|

cct3
Genomic Map