Assay Details
| Gene Symbol: vax2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194409.1 | NP_919390.1 | 2 - 3 | 438 | 107 | |
| GenBank mRNA | AY183363.1 | - | 2 - 3 | 438 | 107 | |
| BC163223.1 | - | 2 - 3 | 530 | 107 | ||
| BC163225.1 | - | 2 - 3 | 530 | 107 | ||
More Information
Set Membership: |
|

vax2
Genomic Map