Assay Details
| Gene Symbol: wtap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199853.1 | NP_956147.1 | 7 - 8 | 777 | 73 | |
| XM_005160635.3 | XP_005160692.1 | 2 - 3 | 879 | 73 | ||
| XM_005160636.3 | XP_005160693.1 | 2 - 3 | 867 | 73 | ||
| GenBank mRNA | BC044427.1 | - | 7 - 8 | 796 | 73 | |
| BC055544.1 | - | 7 - 8 | 777 | 73 | ||
More Information
Set Membership: |
|

wtap
Genomic Map