Assay Details
| Gene Symbol: rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199855.1 | NP_956149.1 | 2 - 3 | 475 | 64 | |
| XM_021481347.1 | XP_021337022.1 | 2 - 3 | 486 | 64 | ||
| GenBank mRNA | BC053301.1 | - | 2 - 3 | 475 | 64 | |
More Information
Set Membership: |
|

rnf168
Genomic Map