Assay Details
| Gene Symbol: umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200174.2 | NP_956468.2 | 3 - 4 | 1057 | 65 | |
| GenBank mRNA | BC045440.1 | - | 3 - 4 | 1003 | 65 | |
| BC055000.1 | - | 3 - 4 | 1003 | 65 | ||
More Information
Set Membership: |
|

umps
Genomic Map