Assay Details
| Gene Symbol: wdr77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200534.1 | NP_956828.1 | 344 | 101 | ||
| GenBank mRNA | BC056278.1 | - | 344 | 101 | ||
| BC066433.1 | - | 304 | 101 | |||
More Information
Set Membership: |
|

wdr77
Genomic Map