Assay Details
| Gene Symbol: cct8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201062.1 | NP_957356.1 | 1 - 2 | 132 | 64 | |
| XM_005155366.3 | XP_005155423.1 | 1 - 2 | 162 | 64 | ||
| GenBank mRNA | AY391413.1 | - | 1 - 2 | 148 | 64 | |
| BC050492.1 | - | 1 - 2 | 132 | 64 | ||
| BC065612.1 | - | 1 - 2 | 117 | 64 | ||
| BC164659.1 | - | 1 - 2 | 92 | 64 | ||
More Information
Set Membership: |
|

cct8
Genomic Map