Assay Details
| Gene Symbol: atg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009914.2 | NP_001009914.2 | 6 - 7 | 758 | 67 | |
| NM_205618.2 | NP_991181.2 | 6 - 7 | 705 | 67 | ||
| GenBank mRNA | AY398310.1 | - | 6 - 7 | 663 | 67 | |
| BC077086.1 | - | 6 - 7 | 757 | 67 | ||
| HQ450378.1 | - | 5 - 6 | 574 | 67 | ||
More Information
Set Membership: |
|

atg5
Genomic Map