Assay Details
| Gene Symbol: sh3bp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316330.1 | NP_001303259.1 | 2 - 3 | 308 | 80 | |
| XM_005161697.4 | XP_005161754.1 | 2 - 3 | 322 | 80 | ||
| GenBank mRNA | AB104885.1 | - | 2 - 3 | 267 | 80 | |
| AB114633.1 | - | 2 - 3 | 267 | 80 | ||
| BC063746.1 | - | 2 - 3 | 308 | 80 | ||
| BC097028.1 | - | 2 - 3 | 273 | 80 | ||
More Information
Set Membership: |
|

sh3bp4a
Genomic Map