Assay Details
| Gene Symbol: sh3bp4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316330.1 | NP_001303259.1 | 4 - 5 | 2855 | 61 | |
| XM_005161697.4 | XP_005161754.1 | 4 - 5 | 2869 | 61 | ||
| GenBank mRNA | AB114633.1 | - | 5 - 6 | 2727 | 61 | |
More Information
Set Membership: |
|

sh3bp4a
Genomic Map