Assay Details
| Gene Symbol: cct5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212613.1 | NP_997778.1 | 6 - 7 | 982 | 64 | |
| GenBank mRNA | AY398321.1 | - | 6 - 7 | 982 | 64 | |
| AY648807.1 | - | 6 - 7 | 957 | 64 | ||
| BC068037.1 | - | 6 - 7 | 967 | 64 | ||
More Information
Set Membership: |
|

cct5
Genomic Map