Assay Details
| Gene Symbol: cd82a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212661.2 | NP_997826.2 | 3 - 4 | 243 | 93 | |
| XM_021477403.1 | XP_021333078.1 | 3 - 4 | 281 | 93 | ||
| GenBank mRNA | BC060930.1 | - | 3 - 4 | 207 | 93 | |
| BC063988.1 | - | 3 - 4 | 220 | 93 | ||
| EH583033.1 | - | 3 - 4 | 267 | 93 | ||
More Information
Set Membership: |
|

cd82a
Genomic Map