Assay Details
| Gene Symbol: ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213093.1 | NP_998258.1 | 7 - 8 | 859 | 108 | |
| GenBank mRNA | BC056559.1 | - | 7 - 8 | 871 | 108 | |
| BC066475.1 | - | 7 - 8 | 859 | 108 | ||
More Information
Set Membership: |
|

ddost
Genomic Map