Assay Details
| Gene Symbol: aip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214712.1 | NP_999877.1 | 4 - 5 | 653 | 86 | |
| GenBank mRNA | AF274877.1 | - | 4 - 5 | 653 | 86 | |
| BC078231.1 | - | 4 - 5 | 677 | 86 | ||
More Information
Set Membership: |
|

aip
Genomic Map