Assay Details
| Gene Symbol: akr1b1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002048.1 | NP_001002048.1 | 4 - 5 | 501 | 59 | |
| GenBank mRNA | BC071313.1 | - | 4 - 5 | 501 | 59 | |
More Information
Set Membership: |
|

akr1b1.1
Genomic Map