Assay Details
| Gene Symbol: ebp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002328.1 | NP_001002328.1 | 3 - 4 | 450 | 96 | |
| GenBank mRNA | BC076453.1 | - | 3 - 4 | 450 | 96 | |
| BC163987.1 | - | 2 - 3 | 378 | 96 | ||
More Information
Set Membership: |
|

ebp
Genomic Map