Assay Details
| Gene Symbol: fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003600.2 | NP_001003600.1 | 2 - 3 | 1133 | 142 | |
| XM_005169513.4 | XP_005169570.1 | 2 - 3 | 1159 | 142 | ||
| GenBank mRNA | BC077144.1 | - | 2 - 3 | 1134 | 142 | |
More Information
Set Membership: |
|

fam120b
Genomic Map