Assay Details
| Gene Symbol: erbb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005320.1 | NP_001005320.1 | 24 - 25 | 3307 | 71 | |
| XM_005165701.4 | XP_005165758.1 | 28 - 29 | 4087 | 71 | ||
| XM_005165702.4 | XP_005165759.1 | 27 - 28 | 4069 | 71 | ||
More Information
Set Membership: |
|

erbb3a
Genomic Map