Assay Details
| Gene Symbol: erbb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005320.1 | NP_001005320.1 | 25 - 26 | 3678 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

erbb3a| Gene Symbol: erbb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005320.1 | NP_001005320.1 | 25 - 26 | 3678 | 86 | |
Set Membership: |
|