Assay Details
| Gene Symbol: erbb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005320.1 | NP_001005320.1 | 14 - 15 | 1900 | 56 | |
| XM_005165701.4 | XP_005165758.1 | 18 - 19 | 2680 | 56 | ||
| XM_005165702.4 | XP_005165759.1 | 17 - 18 | 2662 | 56 | ||
More Information
Set Membership: |
|

erbb3a
Genomic Map