Assay Details
| Gene Symbol: erbb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005320.1 | NP_001005320.1 | 18 - 19 | 2464 | 93 | |
| XM_005165701.4 | XP_005165758.1 | 22 - 23 | 3244 | 93 | ||
| XM_005165702.4 | XP_005165759.1 | 21 - 22 | 3226 | 93 | ||
More Information
Set Membership: |
|

erbb3a
Genomic Map