Assay Details
| Gene Symbol: chga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006059.2 | NP_001006059.2 | 3 - 4 | 395 | 64 | |
| XM_005160561.4 | XP_005160618.1 | 3 - 4 | 416 | 64 | ||
| GenBank mRNA | BC083265.1 | - | 3 - 4 | 378 | 64 | |
| BC164760.1 | - | 3 - 4 | 226 | 64 | ||
| DT865424.1 | - | 3 - 4 | 393 | 64 | ||
| KM655711.1 | - | 3 - 4 | 216 | 64 | ||
More Information
Set Membership: |
|

chga
Genomic Map