Assay Details
| Gene Symbol: ptp4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007775.2 | NP_001007776.2 | 5 - 6 | 1026 | 122 | |
| XM_021479604.1 | XP_021335279.1 | 6 - 7 | 814 | 122 | ||
| GenBank mRNA | BC085425.1 | - | 5 - 6 | 122 | ||
| BC164889.1 | - | 4 - 5 | 122 | |||
| EH576864.1 | - | 1 - 2 | 507 | 122 | ||
More Information
Set Membership: |
|

ptp4a1
Genomic Map