Assay Details
| Gene Symbol: gnpda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017867.1 | NP_001017867.1 | 5 - 6 | 663 | 82 | |
| GenBank mRNA | BC092982.1 | - | 5 - 6 | 663 | 82 | |
More Information
Set Membership: |
|

gnpda1
Genomic Map